This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other VRK3 SIRNA Assays ›
Gene Symbol:
VRK3
Gene Name:
vaccinia related kinase 3
Gene Aliases:
-
Chromosome Location:
Chr.19: 49976467 - 50025954 on Build GRCh38
UniGene ID:
Hs.443330
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS181931
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: VRK3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025778.1 NP_001020949.1 9, 10 1167
NM_001308420.1 NP_001295349.1 10, 11 1155
NM_016440.3 NP_057524.3 10, 11 1317
XM_005258971.3 XP_005259028.1 10, 11 1644
XM_005258972.3 XP_005259029.1 9, 10 1494
XM_006723237.3 XP_006723300.1 10, 11 1458
XM_006723238.3 XP_006723301.1 9, 10 1108
XM_011527023.2 XP_011525325.1 10, 11 1644
GenBank mRNA AB031052.1 - 10, 11 1089
AF514788.1 - 10, 11 1149
AK225154.1 - 10, 11 1062
AK292918.1 - 10, 11 1060
AK294802.1 - 7, 8 757
AK303010.1 - 9, 10 942
AK303529.1 - 9, 10 993
AK311220.1 - 11, 12 1120
BC023556.2 - 10, 11 1096 4298578
BC095449.1 - 10, 11 1069 30721912

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of cell shape
peptidyl-serine phosphorylation
positive regulation of phosphoprotein phosphatase activity
negative regulation of ERK1 and ERK2 cascade
protein serine/threonine kinase activity
protein binding
ATP binding
protein phosphatase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0