This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CKLF SIRNA Assays ›
Gene Symbol:
CKLF CKLF-CMTM1
Gene Name:
chemokine-like factor
CKLF-CMTM1 readthrough
Gene Aliases:
C32, CKLF1, CKLF2, CKLF3, CKLF4, HSPC224, UCK-1
-
Chromosome Location:
Chr.16: 66552563 - 66566287 on Build GRCh38
Chromosome Location:
Chr.16: 66552563 - 66579135 on Build GRCh38
UniGene ID:
Hs.15159
Hs.698621
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID HSS181911
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CKLF
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040138.2 NP_001035228.1 2 286
NM_016951.3 NP_058647.1 2 286
NM_181641.2 NP_857592.1 2 286
GenBank mRNA AF057306.1 - 2 264
AF135380.2 - 2 280
AF145216.2 - 2 280
AF151058.1 - 2 283
AK301240.1 - 2 277
AY358999.1 - 2 263
BC004380.1 - 2 269 3639550
DA804889.1 - 2 267
Gene Symbol: CKLF-CMTM1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204099.1 NP_001191028.1 2 286
GenBank mRNA FY212695.1 - 2 170

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell proliferation
neutrophil chemotaxis
secretion by cell
macrophage chemotaxis
lymphocyte chemotaxis
chemokine activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0