This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C9ORF129 SIRNA Assays ›
Gene Symbol:
C9orf129 WNK2
Gene Name:
chromosome 9 open reading frame 129
WNK lysine deficient protein kinase 2
Gene Aliases:
bA165J3.3
NY-CO-43, P/OKcl.13, PRKWNK2, SDCCAG43
Chromosome Location:
Chr.9: 93318199 - 93346414 on Build GRCh38
Chromosome Location:
Chr.9: 93184156 - 93327581 on Build GRCh38
UniGene ID:
Hs.575636
Hs.654856
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID HSS181348
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C9orf129
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098808.1 NP_001092278.1 4 757
XM_006717117.3 XP_006717180.1 3 776
Gene Symbol: WNK2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282394.1 NP_001269323.1 30 8010
XM_005252137.3 XP_005252194.1 30 8131
XM_005252140.2 XP_005252197.1 30 7976
XM_011518926.2 XP_011517228.1 31 8210
XM_011518927.2 XP_011517229.1 31 8207
XM_011518928.2 XP_011517230.1 31 8156
XM_011518929.2 XP_011517231.1 30 8111
XM_011518930.2 XP_011517232.1 31 8075
XM_011518931.2 XP_011517233.1 29 8066
XM_011518932.2 XP_011517234.1 31 8055
XM_011518935.2 XP_011517237.1 29 7919
XM_011518936.2 XP_011517238.1 28 7874
XM_011518937.2 XP_011517239.1 30 7518
XM_011518939.2 XP_011517241.1 30 7563
XM_011518940.2 XP_011517242.1 30 7565
XM_017015045.1 XP_016870534.1 30 8128
XM_017015046.1 XP_016870535.1 31 8072
XM_017015047.1 XP_016870536.1 30 8077
XM_017015048.1 XP_016870537.1 30 8074
XM_017015049.1 XP_016870538.1 29 8032
XM_017015050.1 XP_016870539.1 29 8029
XM_017015051.1 XP_016870540.1 30 7996
XM_017015052.1 XP_016870541.1 29 7978
XM_017015053.1 XP_016870542.1 29 7975
XM_017015054.1 XP_016870543.1 28 7840
XM_017015055.1 XP_016870544.1 28 7837
GenBank mRNA AB051547.2 - 29 7731

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
negative regulation of cell proliferation
intracellular signal transduction
protein autophosphorylation
negative regulation of ERK1 and ERK2 cascade
regulation of ion homeostasis
positive regulation of sodium ion transmembrane transporter activity
protein serine/threonine kinase activity
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0