This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C6ORF222 SIRNA Assays ›
Gene Symbol:
C6orf222
Gene Name:
chromosome 6 open reading frame 222
Gene Aliases:
-
Chromosome Location:
Chr.6: 36315757 - 36337015 on Build GRCh38
UniGene ID:
Hs.162104
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS180411
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C6orf222
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010903.4 NP_001010903.3 11, 12 2096
XM_011514596.2 XP_011512898.1 11, 12 2949
XM_011514597.2 XP_011512899.1 11, 12 2223
XM_011514598.2 XP_011512900.1 10, 11 1606
GenBank mRNA BC140873.1 - 11, 12 2019 9021741
BX648558.1 - 11, 12 2097

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0