This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KEL SIRNA Assays ›
Gene Symbol:
KEL
Gene Name:
Kell blood group, metallo-endopeptidase
Gene Aliases:
CD238, ECE3
Chromosome Location:
Chr.7: 142941114 - 142962416 on Build GRCh38
UniGene ID:
Hs.368588
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS180175
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KEL
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000420.2 NP_000411.1 8 1008
XM_005249993.2 XP_005250050.1 8 859
XM_005249994.3 XP_005250051.1 2 159
GenBank mRNA AK314831.1 - 8 909
BC003135.1 - 8 1008 2906155
BC050639.1 - 8 903 5742350
FN998902.1 - 7 798
HE572750.1 - 7 798
M64934.2 - 8 918

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
cellular calcium ion homeostasis
regulation of cell size
cellular magnesium ion homeostasis
regulation of axon diameter
vasoconstriction
myelination
skeletal muscle fiber development
negative regulation of potassium ion transmembrane transport
metalloendopeptidase activity
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header