This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM111B SIRNA Assays ›
Gene Symbol:
FAM111B
Gene Name:
family with sequence similarity 111 member B
Gene Aliases:
CANP, POIKTMP
Chromosome Location:
Chr.11: 59107185 - 59127415 on Build GRCh38
UniGene ID:
Hs.186579
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS180029
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM111B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142703.1 NP_001136175.1 3 1546
NM_001142704.1 NP_001136176.1 2 1501
NM_198947.3 NP_945185.1 4 1713
GenBank mRNA AK303520.1 - 3 1495
AK304258.1 - 4 1661
AY457926.1 - 4 1662
BC130513.1 - 3 1603 40146544
BC130539.1 - 3 1603 40146570

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0