This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF445 SIRNA Assays ›
Gene Symbol:
ZNF445
Gene Name:
zinc finger protein 445
Gene Aliases:
ZKSCAN15, ZNF168, ZSCAN47
Chromosome Location:
Chr.3: 44439770 - 44477670 on Build GRCh38
UniGene ID:
Hs.250481
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS179859
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF445
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181489.5 NP_852466.1 3 522
XM_005265102.2 XP_005265159.1 2 380
XM_011533674.2 XP_011531976.1 2 345
XM_017006345.1 XP_016861834.1 2 437
XM_017006346.1 XP_016861835.1 2 380
GenBank mRNA AK310588.1 - 2 381
AY262260.1 - 1 325
AY279350.1 - 2 341
AY295873.2 - 3 522
BC101486.1 - 1 236 8068992
BC143439.1 - 1 237 9051947
L32162.1 - 2 341

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0