This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HOXD9 SIRNA Assays ›
Gene Symbol:
HOXD9
Gene Name:
homeobox D9
Gene Aliases:
HOX4, HOX4C, Hox-4.3, Hox-5.2
Chromosome Location:
Chr.2: 176122685 - 176124917 on Build GRCh38
UniGene ID:
Hs.236646
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS179344
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HOXD9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014213.3 NP_055028.3 2 1037
GenBank mRNA BC044855.1 - 2 1037 5191006
X59372.1 - 2 959

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
single fertilization
skeletal muscle tissue development
adult locomotory behavior
anterior/posterior pattern specification
proximal/distal pattern formation
mammary gland development
embryonic forelimb morphogenesis
hindlimb morphogenesis
positive regulation of transcription from RNA polymerase II promoter
embryonic skeletal system morphogenesis
peripheral nervous system neuron development
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0