This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GPS1 SIRNA Assays ›
Gene Symbol:
GPS1
Gene Name:
G protein pathway suppressor 1
Gene Aliases:
COPS1, CSN1, SGN1
Chromosome Location:
Chr.17: 82051477 - 82057470 on Build GRCh38
UniGene ID:
Hs.268530
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS178901
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GPS1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321089.1 NP_001308018.1 4 529
NM_001321090.1 NP_001308019.1 4 529
NM_001321091.1 NP_001308020.1 4 529
NM_001321092.1 NP_001308021.1 3 325
NM_001321093.1 NP_001308022.1 4 410
NM_004127.5 NP_004118.3 3 325
NM_212492.2 NP_997657.1 3 820
XM_005256350.1 XP_005256407.1 4 497
XM_005256353.1 XP_005256410.1 4 497
XM_005256359.1 XP_005256416.1 3 289
XM_005256360.3 XP_005256417.1 3 330
XM_011523569.1 XP_011521871.1 4 350
XM_011523570.1 XP_011521872.1 4 400
XM_017024524.1 XP_016880013.1 3 307
XM_017024525.1 XP_016880014.1 4 497
XM_017024526.1 XP_016880015.1 3 307
XM_017024527.1 XP_016880016.1 3 330
XM_017024528.1 XP_016880017.1 3 273
XM_017024529.1 XP_016880018.1 3 273
XM_017024530.1 XP_016880019.1 3 273
XM_017024531.1 XP_016880020.1 4 350
XM_017024532.1 XP_016880021.1 3 289
XM_017024533.1 XP_016880022.1 3 317
XM_017024534.1 XP_016880023.1 4 350
XM_017024535.1 XP_016880024.1 4 350
GenBank mRNA AB209596.1 - 2 929
AK026610.1 - 4 372
AK222508.1 - 3 290
AK225248.1 - 3 287
AK289435.1 - 3 443
AK297872.1 - 3 301
BC000155.2 - 3 281 2900944
BC064503.1 - 3 732 6572943
BG775520.1 - 4 388
BP306851.1 - 3 325
BT009834.1 - 2 210
U20285.2 - 3 317

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

inactivation of MAPK activity
nucleotide-excision repair, DNA damage recognition
transcription-coupled nucleotide-excision repair
cell cycle
JNK cascade
COP9 signalosome assembly
cullin deneddylation
negative regulation of GTPase activity
GTPase inhibitor activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0