This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM171A2 SIRNA Assays ›
Gene Symbol:
FAM171A2
Gene Name:
family with sequence similarity 171 member A2
Gene Aliases:
-
Chromosome Location:
Chr.17: 44353215 - 44363867 on Build GRCh38
UniGene ID:
Hs.154001
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS178640
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM171A2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198475.2 NP_940877.2 5 802
XM_017024490.1 XP_016879979.1 3 369
GenBank mRNA BC029298.1 - 5 818
BC030200.1 - 5 802

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0