This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PCNX1 SIRNA Assays ›
Gene Symbol:
PCNX1
Gene Name:
pecanex homolog 1 (Drosophila)
Gene Aliases:
PCNX, PCNXL1, pecanex
Chromosome Location:
Chr.14: 70907405 - 71115382 on Build GRCh38
UniGene ID:
Hs.446559
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS177071
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PCNX1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308160.1 NP_001295089.1 17 3929
NM_014982.2 NP_055797.2 19 4262
XM_005267420.4 XP_005267477.1 19 4224
XM_005267421.4 XP_005267478.1 19 4221
XM_005267422.4 XP_005267479.1 19 4209
XM_005267423.4 XP_005267480.1 19 4209
XM_005267424.3 XP_005267481.2 19 3928
XM_005267425.4 XP_005267482.1 17 3906
XM_005267427.4 XP_005267484.1 19 4224
XM_005267428.3 XP_005267485.1 12 1212
XM_006720082.3 XP_006720145.1 10 1151
XM_017021096.1 XP_016876585.1 17 3906
XM_017021097.1 XP_016876586.1 12 1476
GenBank mRNA AB018348.1 - 9 992
AF233450.1 - 19 4262
BC140784.1 - 17 3595 9021652

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0