This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EYA2 SIRNA Assays ›
Gene Symbol:
EYA2
Gene Name:
EYA transcriptional coactivator and phosphatase 2
Gene Aliases:
EAB1
Chromosome Location:
Chr.20: 46894624 - 47188844 on Build GRCh38
UniGene ID:
Hs.472877
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS176650
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: EYA2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005244.4 NP_005235.3 11 1372
NM_172110.3 NP_742108.2 11 1372
XM_005260327.2 XP_005260384.1 9 819
XM_017027721.1 XP_016883210.1 10 1000
GenBank mRNA AF055015.1 - 7 686
AK093582.1 - 2 1256
AK293032.1 - 11 1076
AY705349.1 - 9 918
BC000289.2 - 5 524
BC008803.2 - 11 1054 3944547
BC013882.2 - 11 1372
BT006682.1 - 10 926
U71207.1 - 10 1000
U81601.1 - 2 116

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
mesodermal cell fate specification
striated muscle tissue development
histone dephosphorylation
peptidyl-tyrosine dephosphorylation
extrinsic apoptotic signaling pathway in absence of ligand
mitochondrial outer membrane permeabilization
magnesium ion binding
protein tyrosine phosphatase activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0