This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KBTBD12 SIRNA Assays ›
Gene Symbol:
KBTBD12
Gene Name:
kelch repeat and BTB domain containing 12
Gene Aliases:
KLHDC6
Chromosome Location:
Chr.3: 127915232 - 127987858 on Build GRCh38
UniGene ID:
Hs.132087
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS176069
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KBTBD12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207335.2 NP_997218.2 1 193
XM_005247167.1 XP_005247224.1 2 657
XM_005247168.4 XP_005247225.1 2 735
XM_011512490.2 XP_011510792.1 2 657
XM_011512491.2 XP_011510793.1 2 657
XM_011512492.2 XP_011510794.1 2 657

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
ubiquitin-protein transferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0