This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF38 SIRNA Assays ›
Gene Symbol:
RNF38
Gene Name:
ring finger protein 38
Gene Aliases:
-
Chromosome Location:
Chr.9: 36336398 - 36487384 on Build GRCh38
UniGene ID:
Hs.333503
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS175709
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RNF38
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022781.4 NP_073618.3 11 1621
NM_194328.2 NP_919309.1 11 1788
NM_194329.2 NP_919310.1 10 1471
NM_194330.2 NP_919311.1 11 1519
NM_194332.2 NP_919313.1 10 1638
XM_005251366.3 XP_005251423.1 11 1625
XM_005251367.3 XP_005251424.1 11 1523
XM_005251368.3 XP_005251425.1 11 1637
XM_006716721.3 XP_006716784.1 10 1485
XM_011517712.2 XP_011516014.1 12 1561
XM_011517713.2 XP_011516015.1 11 1625
XM_017014294.1 XP_016869783.1 10 1542
XM_017014295.1 XP_016869784.1 11 1652
XM_017014296.1 XP_016869785.1 10 1222
XM_017014297.1 XP_016869786.1 10 1218
GenBank mRNA AF394047.1 - 11 1743
AK024996.1 - 9 1131
AK093480.1 - 11 1687
AK097653.1 - 11 1680
AL136817.1 - 11 1540
AX748155.1 - 11 1687
BC033786.1 - 10 1471 5166325

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
male gonad development
protein ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
zinc ion binding
ligase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0