This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LRRC39 SIRNA Assays ›
Gene Symbol:
LRRC39
Gene Name:
leucine rich repeat containing 39
Gene Aliases:
-
Chromosome Location:
Chr.1: 100148448 - 100178273 on Build GRCh38
UniGene ID:
Hs.44277
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS174849
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LRRC39
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256385.1 NP_001243314.1 8 955
NM_001256386.1 NP_001243315.1 8 955
NM_001256387.1 NP_001243316.1 7 898
NM_144620.3 NP_653221.1 8 938
GenBank mRNA AK096892.1 - 8 881
AL833417.1 - 8 903
AY358131.1 - 7 871
BC009613.1 - 8 880 4245944

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header