This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF573 SIRNA Assays ›
Gene Symbol:
ZNF573
Gene Name:
zinc finger protein 573
Gene Aliases:
-
Chromosome Location:
Chr.19: 37738274 - 37779626 on Build GRCh38
UniGene ID:
Hs.531262
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS174788
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF573
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172689.1 NP_001166160.1 5, 6 565
NM_001172690.1 NP_001166161.1 4, 5 374
NM_001172691.1 NP_001166162.1 4, 5 368
NM_001172692.1 NP_001166163.1 3, 4 241
NM_152360.3 NP_689573.3 7, 8 633
XM_011526446.2 XP_011524748.1 5, 6 452
XM_017026275.1 XP_016881764.1 5, 6 559
XM_017026276.1 XP_016881765.1 5, 6 1281
XM_017026277.1 XP_016881766.1 5, 6 446
XM_017026278.1 XP_016881767.1 3, 4 319
XM_017026279.1 XP_016881768.1 3, 4 264
XM_017026280.1 XP_016881769.1 4, 5 510
XM_017026281.1 XP_016881770.1 3, 4 1103
GenBank mRNA AK055483.1 - 2, 3 383
AK074539.1 - 5, 6 561
BC029468.2 - 322
BC042170.2 - 3, 4 328 5014852
BC064962.1 - 7, 8 603 6141400
BG572864.1 - 4, 5 352
BP274132.1 - 4, 5 370
DB121870.1 - 3, 4 214

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0