This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF573 SIRNA Assays ›
Gene Symbol:
ZNF573
Gene Name:
zinc finger protein 573
Gene Aliases:
-
Chromosome Location:
Chr.19: 37738274 - 37779626 on Build GRCh38
UniGene ID:
Hs.531262
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS174787
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF573
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172689.1 NP_001166160.1 6 1534
NM_001172690.1 NP_001166161.1 5 1343
NM_001172691.1 NP_001166162.1 5 1337
NM_001172692.1 NP_001166163.1 4 1210
NM_152360.3 NP_689573.3 8 1602
XM_011526446.2 XP_011524748.1 6 1421
XM_017026275.1 XP_016881764.1 6 1528
XM_017026276.1 XP_016881765.1 6 2250
XM_017026277.1 XP_016881766.1 6 1415
XM_017026278.1 XP_016881767.1 4 1288
XM_017026279.1 XP_016881768.1 4 1233
XM_017026280.1 XP_016881769.1 5 1479
XM_017026281.1 XP_016881770.1 4 2072
GenBank mRNA AK055483.1 - 3 1352
AK074539.1 - 6 1530
BC015418.1 - 1 589
BC042170.2 - 4 1297 5014852
BC051263.1 - 1 164
BC064962.1 - 8 1572 6141400

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0