This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM219 SIRNA Assays ›
Gene Symbol:
TMEM219
Gene Name:
transmembrane protein 219
Gene Aliases:
IGFBP-3R
Chromosome Location:
Chr.16: 29962030 - 29973052 on Build GRCh38
UniGene ID:
Hs.460574
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS174715
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM219
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083613.1 NP_001077082.1 3 397
NM_194280.3 NP_919256.1 3 394
XM_005255123.1 XP_005255180.1 3 396
GenBank mRNA BC017488.1 - 2 162
BC031217.2 - 2 291
BC050051.1 - 3 424 5278133
BF724791.1 - 3 354
BU857546.1 - 362
CF453990.1 - 341
DB506492.1 - 3 397
FJ748884.1 - 2 257

Back To Top

More Information


Gene Ontology Categories:

Function(s)

apoptotic process
regulation of apoptotic process

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0