This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ANO4 SIRNA Assays ›
Gene Symbol:
ANO4
Gene Name:
anoctamin 4
Gene Aliases:
TMEM16D
Chromosome Location:
Chr.12: 100717457 - 101128641 on Build GRCh38
UniGene ID:
Hs.58785
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS174644
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ANO4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286615.1 NP_001273544.1 25 3057
NM_001286616.1 NP_001273545.1 24 2625
NM_178826.3 NP_849148.2 24 2952
XM_011537911.2 XP_011536213.1 27 3052
XM_011537912.2 XP_011536214.1 27 3052
XM_011537913.2 XP_011536215.1 26 3007
XM_011537914.2 XP_011536216.1 24 2563
XM_011537915.2 XP_011536217.1 25 2875
XM_011537916.2 XP_011536218.1 25 2716
XM_017018808.1 XP_016874297.1 24 2563
XM_017018809.1 XP_016874298.1 26 3143
XM_017018810.1 XP_016874299.1 24 2805
XM_017018811.1 XP_016874300.1 23 2760
XM_017018812.1 XP_016874301.1 23 4778
XM_017018813.1 XP_016874302.1 22 2628
XM_017018814.1 XP_016874303.1 25 5017
XM_017018815.1 XP_016874304.1 25 5017
XM_017018816.1 XP_016874305.1 25 3100
XM_017018817.1 XP_016874306.1 25 5052
GenBank mRNA AK091540.1 - 15 1573
AK091591.1 - 24 2741
AK092596.1 - 12 1139
AK293810.1 - 25 2695
AK316105.1 - 25 2696
BC109308.2 - 24 2610 40035759

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chloride transport
lipid transport
ion transmembrane transport
calcium activated phosphatidylserine scrambling
calcium activated phosphatidylcholine scrambling
calcium activated galactosylceramide scrambling
chloride transmembrane transport
intracellular calcium activated chloride channel activity
phospholipid scramblase activity
protein dimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0