This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CMTM1 SIRNA Assays ›
Gene Symbol:
CMTM1
Gene Name:
CKLF like MARVEL transmembrane domain containing 1
Gene Aliases:
CKLFH, CKLFH1, CKLFSF1
Chromosome Location:
Chr.16: 66566391 - 66579135 on Build GRCh38
UniGene ID:
Hs.15159
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS174296
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CMTM1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052999.3 NP_443725.3 2 629
NM_181268.2 NP_851785.2 2 629
NM_181269.2 NP_851786.1 2 334
NM_181270.2 NP_851787.1 2 334
GenBank mRNA AF278576.1 - 2 249
AF278577.1 - 2 249
AY174118.1 - 2 338
AY174119.1 - 2 338
AY174125.1 - 2 338
AY174126.1 - 2 338
AY174129.1 - 2 164
AY174134.1 - 2 329
AY174135.1 - 2 329
AY174136.1 - 2 329
AY174137.1 - 2 329
AY174138.1 - 2 329
BC057852.1 - 2 629 5273108
DQ309766.1 - 2 329

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chemotaxis
cytokine activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header