This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RAB35 SIRNA Assays ›
Gene Symbol:
RAB35
Gene Name:
RAB35, member RAS oncogene family
Gene Aliases:
H-ray, RAB1C, RAY
Chromosome Location:
Chr.12: 120095094 - 120116839 on Build GRCh38
UniGene ID:
Hs.524788
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS174045
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RAB35
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167606.1 NP_001161078.1 4 492
NM_006861.6 NP_006852.1 4 492
XM_011537779.1 XP_011536081.1 4 429
XM_017018723.1 XP_016874212.1 4 394
GenBank mRNA AF498960.1 - 4 303
AK125543.1 - 5 1267
AK298391.1 - 1 12
AK304620.1 - 4 399
AK312538.1 - 4 402
BC015931.2 - 4 407 3907209
BM467132.1 - 4 474
BQ233709.1 - 4 448
BT020024.1 - 4 303
CR536486.1 - 4 303
CR541683.1 - 4 303
DC349830.1 - 4 489
DC370610.1 - 4 492
DC408044.1 - 4 492
X79781.1 - 4 384

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cytokinesis
ER to Golgi vesicle-mediated transport
small GTPase mediated signal transduction
protein localization
protein transport
endosomal transport
antigen processing and presentation
neuron projection development
endocytic recycling
protein localization to endosome
plasma membrane to endosome transport
cellular response to nerve growth factor stimulus
GTPase activity
protein binding
GTP binding
phosphatidylinositol-4,5-bisphosphate binding
GDP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header