This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF197 SIRNA Assays ›
Gene Symbol:
ZNF197
Gene Name:
zinc finger protein 197
Gene Aliases:
D3S1363E, P18, VHLaK, ZKSCAN9, ZNF166, ZNF20, ZSCAN41
Chromosome Location:
Chr.3: 44625017 - 44648471 on Build GRCh38
UniGene ID:
Hs.157035
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS173445
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF197
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005264783.4 XP_005264840.1 5 905
XM_017005495.1 XP_016860984.1 5 877
GenBank mRNA AF011573.1 - 4 692
AK314196.1 - 5 873
AY074878.1 - 5 873
AY261677.1 - 4 706
AY279353.1 - 5 894
BC017584.1 - 4 858
Z21707.1 - 4 604

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0