This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM262 SIRNA Assays ›
Gene Symbol:
TMEM262
Gene Name:
transmembrane protein 262
Gene Aliases:
-
Chromosome Location:
Chr.11: 65088372 - 65089109 on Build GRCh38
UniGene ID:
Hs.712317
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS171057
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM262
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242631.2 NP_001229560.1 1 146
NM_001282448.1 NP_001269377.1 1 146
GenBank mRNA AI015611.1 - 3 334
AK124141.1 - 1 144
BX093670.1 - 1 101
HY032040.1 - 1 148

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0