This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CISD2 SIRNA Assays ›
Gene Symbol:
CISD2
Gene Name:
CDGSH iron sulfur domain 2
Gene Aliases:
ERIS, Miner1, NAF-1, WFS2, ZCD2
Chromosome Location:
Chr.4: 102868978 - 102892807 on Build GRCh38
UniGene ID:
Hs.444955
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS166719
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CISD2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008388.4 NP_001008389.1 1 143
GenBank mRNA AK292134.1 - 1 80
BC032300.1 - 1 80 5105935
BX537971.1 - 1 81
DN993861.1 - 1 148

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitophagy
multicellular organism aging
regulation of autophagy
protein binding
protein homodimerization activity
poly(A) RNA binding
metal ion binding
2 iron, 2 sulfur cluster binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header