siRNA Details
Gene Symbol: LINC00674 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027418.1 | 6 | 10118 | |||
GenBank mRNA | AK091150.1 | - | 1 | 1655 | ||
AK092798.1 | - | 1 | 839 |

Gene Symbol: LINC00674 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027418.1 | 6 | 10118 | |||
GenBank mRNA | AK091150.1 | - | 1 | 1655 | ||
AK092798.1 | - | 1 | 839 |