siRNA Details
Gene Symbol: HERC2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002824.3 | 35 | 5762 | |||
GenBank mRNA | AA976147.1 | - | 1 | 310 | ||
AB002391.2 | - | 32 | 5449 | |||
AF041080.1 | - | LIT2190 HERC2P2 |
36 | 5762 | ||
AK095014.1 | - | 12 | 3045 | |||
CR749409.1 | - | 24 | 4467 |
