This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HERC2P2 SIRNA Assays ›
Gene Symbol:
HERC2P2
Gene Name:
hect domain and RLD 2 pseudogene 2
Gene Aliases:
D15F37S3, MN7
Chromosome Location:
Chr.15: 22494837 - 22590831 on Build GRCh38
UniGene ID:
Hs.458334
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS160303
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HERC2P2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002824.3 34, 35 5182
GenBank mRNA AB002391.2 - 31, 32 4869

Back To Top

More Information


Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0