This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HERC2P9 SIRNA Assays ›
Gene Symbol:
HERC2P9 HERC2P2
Gene Name:
hect domain and RLD 2 pseudogene 9
hect domain and RLD 2 pseudogene 2
Gene Aliases:
-
D15F37S3, MN7
Chromosome Location:
Chr.15: 28654442 - 28685264 on Build GRCh38
Chromosome Location:
Chr.15: 22494837 - 22590831 on Build GRCh38
UniGene ID:
Hs.531509
Hs.458334
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID HSS160302
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HERC2P9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_036443.1 10 1513
GenBank mRNA AK293512.1 - 10 1564
BC047911.1 - 10 1513
Gene Symbol: HERC2P2
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002824.3
32 4888
GenBank mRNA AB002391.2 -
29 4575
AF041080.1 - LIT2190
HERC2P2
32 4894
AF041081.1 - LIT2191
HERC2P3
28 4851
AK095014.1 -
8 2016
CR749409.1 -
21 3330

Back To Top

More Information


Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0