siRNA Details
Gene Symbol: ZNF818P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073396.1 | 1 | 97 | |||
GenBank mRNA | AK128250.1 | - | 1 | 70 | ||
BF908632.1 | - | 1 | 113 |

Gene Symbol: ZNF818P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073396.1 | 1 | 97 | |||
GenBank mRNA | AK128250.1 | - | 1 | 70 | ||
BF908632.1 | - | 1 | 113 |