This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NHLRC3 SIRNA Assays ›
Gene Symbol:
NHLRC3
Gene Name:
NHL repeat containing 3
Gene Aliases:
-
Chromosome Location:
Chr.13: 39038311 - 39050109 on Build GRCh38
UniGene ID:
Hs.507783
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS156994
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NHLRC3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012754.3 NP_001012772.1 1 365
NM_001017370.2 NP_001017370.1 1 365
GenBank mRNA AK299198.1 - 1 278
AK300260.1 - 1 278
AL833329.1 - 1 370
BC140877.1 - 1 291 9021745
BC140878.1 - 1 291 9021746
BX647872.1 - 1 255
BX648122.1 - 1 317
CR749318.1 - 1 286

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0