This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC554249 SIRNA Assays ›
Gene Symbol:
LOC554249 GLIDR KGFLP1 KGFLP2 LOC102724238 LOC403323 LOC728673
Gene Name:
uncharacterized LOC554249
glioblastoma down-regulated RNA
fibroblast growth factor 7 pseudogene
keratinocyte growth factor-like protein 2
uncharacterized LOC102724238
uncharacterized LOC403323
uncharacterized LOC728673
Gene Aliases:
-
LINC01172, TCONS_00015562
-
-
-
-
-
Chromosome Location:
Chr.9: 39873857 - 39875395 on Build GRCh38
Chromosome Location:
Chr.9: 39803496 - 39810159 on Build GRCh38
Chromosome Location:
Chr.9: 62376256 - 62435519 on Build GRCh38
Chromosome Location:
Chr.9: 39813784 - 39874566 on Build GRCh38
Chromosome Location:
Chr.9: 62375328 - 62376870 on Build GRCh38
Chromosome Location:
Chr.9: 62857856 - 62898087 on Build GRCh38
Chromosome Location:
Chr.9: 62897376 - 62899777 on Build GRCh38
UniGene ID:
Hs.721511
Hs.380240
Hs.439341
Hs.536967
Hs.536967
Hs.646340
Hs.380240
Species:
Human
Species Specific ID (Flybase ID):
-
-
-
-
-
-
-
siRNA ID HSS156680
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC554249
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_126049.1 1 84
Gene Symbol: GLIDR
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015363.2 1 144
NR_126044.1 1 144
NR_126045.1 1 144
NR_126046.1 1 144
GenBank mRNA AK092305.1 - 1 43
AK095690.1 - 1 45
BC027338.1 - 1 48
Gene Symbol: KGFLP1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003674.2 1 524
NR_047527.1 1 524
GenBank mRNA AF523265.1 - 1 619
AK054997.1 - 1 263
AK307215.1 - 1 255
AY098593.1 - 1 515
BC101842.1 - 1 524
BC112977.1 - 1 522
BC112978.1 - 1 522
DB501751.1 - 1 65
Gene Symbol: KGFLP2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003670.1 1 623
GenBank mRNA AY098594.1 - 1 515
BC143690.1 - 1 525
Gene Symbol: LOC102724238
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_126048.1 1 88
GenBank mRNA AK290223.1 - 1 531
AK292642.1 - 1 15
Gene Symbol: LOC403323
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_122077.1 1 635
GenBank mRNA AK000451.1 - 1 635
Gene Symbol: LOC728673
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_126047.1 1 74
GenBank mRNA DA480961.1 - 1 36

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0