This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC284798 SIRNA Assays ›
Gene Symbol:
LOC284798
Gene Name:
uncharacterized LOC284798
Gene Aliases:
-
Chromosome Location:
Chr.20: 25140798 - 25148790 on Build GRCh38
UniGene ID:
Hs.334934
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS154712
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC284798
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027091.1 4 1340
NR_027092.1 4 633
NR_027093.1 4 740
GenBank mRNA AK095800.1 - 4 1340
AK309371.1 - 4 633
BC105718.1 - 4 740
BC105722.1 - 4 740

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0