This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GCSAM SIRNA Assays ›
Gene Symbol:
GCSAM
Gene Name:
germinal center associated signaling and motility
Gene Aliases:
GCAT2, GCET2, HGAL
Chromosome Location:
Chr.3: 112120841 - 112133305 on Build GRCh38
UniGene ID:
Hs.49614
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS154085
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GCSAM
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190259.1 NP_001177188.1 6 630
NM_001190260.1 NP_001177189.1 5 579
NM_152785.4 NP_689998.1 6 624
XM_005247272.2 XP_005247329.1 6 1048
XM_011512627.2 XP_011510929.1 6 502
XM_011512628.2 XP_011510930.1 6 559
XM_011512629.1 XP_011510931.1 6 1051
XM_011512630.1 XP_011510932.1 6 1057
GenBank mRNA AF521911.1 - 6 611
AK313368.1 - 6 571
AY212246.1 - 6 545
BC030506.1 - 6 492 4385178
BM456595.1 - 6 580
BP224611.1 - 5 528

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of B cell receptor signaling pathway
negative regulation of lymphocyte migration
actin binding
protein binding
protein kinase binding
myosin II binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0