siRNA Details
Gene Symbol: NBPF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011540763.1 | XP_011539065.1 | 16 | 3176 | ||
GenBank mRNA | AK057395.1 | - | 14 | 2546 |

Gene Symbol: NBPF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011540763.1 | XP_011539065.1 | 16 | 3176 | ||
GenBank mRNA | AK057395.1 | - | 14 | 2546 |