This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MRGPRE SIRNA Assays ›
Gene Symbol:
MRGPRE
Gene Name:
MAS related GPR family member E
Gene Aliases:
GPR167, MGRF, MRGE
Chromosome Location:
Chr.11: 3227811 - 3232386 on Build GRCh38
UniGene ID:
Hs.745512
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS151077
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MRGPRE
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039165.2 NP_001034254.2 2 513
GenBank mRNA AK293940.1 - 2 513
AY255572.1 - 1 35
BC104889.1 - 1 237 8143892
BC112203.1 - 1 249 8327671
DC309261.1 - 2 513

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
positive regulation of protein targeting to mitochondrion
G-protein coupled receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header