This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RMI2 SIRNA Assays ›
Gene Symbol:
RMI2
Gene Name:
RecQ mediated genome instability 2
Gene Aliases:
BLAP18, C16orf75
Chromosome Location:
Chr.16: 11345438 - 11351763 on Build GRCh38
UniGene ID:
Hs.347524
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS151042
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RMI2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152308.2 NP_689521.1 2 1408
NR_130754.1 2 1185
GenBank mRNA AK123764.1 - 2 1397
BC013040.2 - 2 1376 3542171
BC022427.1 - 2 1392 4252688
BC031016.1 - 2 1396 4659915
BC039361.1 - 5 1830 5270594

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

DNA synthesis involved in DNA repair
strand displacement
DNA replication
regulation of signal transduction by p53 class mediator
DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0