This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC102724354 SIRNA Assays ›
Gene Symbol:
LOC102724354 LINC00313
Gene Name:
uncharacterized LOC102724354
long intergenic non-protein coding RNA 313
Gene Aliases:
-
C21orf84, CH507-42P11.5, NCRNA00313
Chromosome Location:
Chr.21: 6059029 - 6076305 on Build GRCh38
Chromosome Location:
Chr.21: 43462094 - 43478223 on Build GRCh38
UniGene ID:
Hs.592161
Hs.592161
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID HSS150891
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC102724354
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_136540.1 2, 3 121
GenBank mRNA AW206028.1 - 3, 4 415
HY162931.1 - 1, 2 79
Gene Symbol: LINC00313
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026863.1 1, 2 160
GenBank mRNA AF426261.1 - 1, 2 160
BC107109.2 - 1, 2 91

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0