This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GGTLC1 SIRNA Assays ›
Gene Symbol:
GGTLC1
Gene Name:
gamma-glutamyltransferase light chain 1
Gene Aliases:
GGTL6, GGTLA3, GGTLA4, dJ831C21.1, dJ831C21.2
Chromosome Location:
Chr.20: 23985053 - 23989134 on Build GRCh38
UniGene ID:
Hs.355394
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS150636
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GGTLC1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178311.2 NP_842563.1 5 658
NM_178312.2 NP_842564.1 5 586
XM_005260863.4 XP_005260920.1 4 1094
XM_005260864.4 XP_005260921.1 5 1247
XM_005260865.4 XP_005260922.1 6 1166
XM_011529391.2 XP_011527693.1 4 661
XM_011529392.2 XP_011527694.1 6 1105
XM_011529393.2 XP_011527695.1 5 952
XM_017028126.1 XP_016883615.1 5 575
GenBank mRNA BC040904.1 - 5 658 5744021
L20491.1 - 5 586
L20492.1 - 4 636

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
leukotriene biosynthetic process
gamma-glutamyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0