This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MIDN SIRNA Assays ›
Gene Symbol:
MIDN
Gene Name:
midnolin
Gene Aliases:
-
Chromosome Location:
Chr.19: 1248552 - 1259143 on Build GRCh38
UniGene ID:
Hs.465529
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS150418
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MIDN
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177401.4 NP_796375.3 5 965
XM_005259671.3 XP_005259728.1 6 1094
XM_005259672.3 XP_005259729.1 6 1095
GenBank mRNA AA421908.1 - 2 133
BC094778.1 - 4 859 30342357

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of glucokinase activity
negative regulation of insulin secretion
kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0