This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZDHHC12 SIRNA Assays ›
Gene Symbol:
ZDHHC12
Gene Name:
zinc finger DHHC-type containing 12
Gene Aliases:
DHHC-12, ZNF400
Chromosome Location:
Chr.9: 128720869 - 128724138 on Build GRCh38
UniGene ID:
Hs.133122
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS150173
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZDHHC12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318015.1 NP_001304944.1 3 489
NM_001318016.1 NP_001304945.1 3 324
NM_001318020.1 NP_001304949.1 3 324
NM_001318023.1 NP_001304952.1 3 324
NM_032799.4 NP_116188.2 3 324
XM_011519116.2 XP_011517418.1 3 498
GenBank mRNA AK027430.1 - 3 317
AK075332.1 - 3 317
AK092843.1 - 2 1741
AK315746.1 - 3 320
BC009280.2 - 3 301 4302257
BC048251.1 - 3 470 5418875
BG767826.1 - 3 298
BM043458.1 - 3 292
BM745076.1 - 3 316
BU506687.1 - 3 325

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein palmitoylation
zinc ion binding
palmitoyltransferase activity
protein-cysteine S-palmitoyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0