This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TNRC18 SIRNA Assays ›
Gene Symbol:
TNRC18
Gene Name:
trinucleotide repeat containing 18
Gene Aliases:
CAGL79, TNRC18A
Chromosome Location:
Chr.7: 5306790 - 5426043 on Build GRCh38
UniGene ID:
Hs.520638
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS150040
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TNRC18
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080495.2 NP_001073964.2 5 2193
XM_017012728.1 XP_016868217.1 5 4690
XM_017012729.1 XP_016868218.1 4 2397
XM_017012730.1 XP_016868219.1 4 2015
XM_017012731.1 XP_016868220.1 4 2016
XM_017012732.1 XP_016868221.1 4 2016
XM_017012733.1 XP_016868222.1 3 1872
XM_017012734.1 XP_016868223.1 4 2898
XM_017012735.1 XP_016868224.1 4 1900
XM_017012736.1 XP_016868225.1 4 2487
XM_017012737.1 XP_016868226.1 4 4260
XM_017012738.1 XP_016868227.1 4 1782
XM_017012739.1 XP_016868228.1 4 2017
XM_017012740.1 XP_016868229.1 4 2017
GenBank mRNA CR993399.1 - 1 50

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chromatin silencing
heterochromatin assembly
transcription regulatory region sequence-specific DNA binding
chromatin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-77574795ff-5hfml:80/100.66.128.142:80.
git-commit: 5cc8a939e1477954742eeb281a87e54f28adfb77
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header