This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C3ORF36 SIRNA Assays ›
Gene Symbol:
C3orf36
Gene Name:
chromosome 3 open reading frame 36
Gene Aliases:
-
Chromosome Location:
Chr.3: 133928145 - 133929812 on Build GRCh38
UniGene ID:
Hs.287691
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS149396
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C3orf36
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025041.2 NP_079317.2 1 1538
GenBank mRNA AK025826.1 - 1 1400
AK225383.1 - 1 1400
BC051274.1 - 1 1538
BC064502.1 - 1 1538
BC082257.1 - 1 1538
BC104161.1 - 1 717 40029559
BC104162.1 - 1 714 40029562

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0