This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NDUFB7 SIRNA Assays ›
Gene Symbol:
NDUFB7
Gene Name:
NADH:ubiquinone oxidoreductase subunit B7
Gene Aliases:
B18, CI-B18
Chromosome Location:
Chr.19: 14566078 - 14572077 on Build GRCh38
UniGene ID:
Hs.532853
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS143068
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NDUFB7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004146.5 NP_004137.2 1, 2 200
XM_011528039.2 XP_011526341.1 1, 2 792
GenBank mRNA AF112200.1 - 1, 2 186
AF217091.1 - 1, 2 145
BC002595.2 - 1, 2 185 3140806
BU565399.1 - 201
CB124371.1 - 1, 2 200
CR450354.1 - 1, 2 123
M33374.1 - 1, 2 158

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitochondrial electron transport, NADH to ubiquinone
mitochondrial respiratory chain complex I assembly
NADH dehydrogenase (ubiquinone) activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0