siRNA Details
Gene Symbol: FLJ35816 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093135.1 | - | 1 | 461 | ||
BC121811.1 | - | 1 | 395 |

Gene Symbol: FLJ35816 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093135.1 | - | 1 | 461 | ||
BC121811.1 | - | 1 | 395 |