This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC400661 SIRNA Assays ›
Gene Symbol:
LOC400661
Gene Name:
uncharacterized LOC400661
Gene Aliases:
-
Chromosome Location:
Chr.18: 76690029 - 76693636 on Build GRCh38
UniGene ID:
Hs.514963
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS140330
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC400661
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_122073.1 1 387
GenBank mRNA AK126829.1 - 1 387
BC119774.1 - 1 327 40109466
BC119776.1 - 1 327 40109460

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0