This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SIMC1 SIRNA Assays ›
Gene Symbol:
SIMC1
Gene Name:
SUMO interacting motifs containing 1
Gene Aliases:
C5orf25, OOMA1, PLEIAD
Chromosome Location:
Chr.5: 176238359 - 176345991 on Build GRCh38
UniGene ID:
Hs.719847
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS139691
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SIMC1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308195.1 NP_001295124.1 5, 6 2049
NM_001308196.1 NP_001295125.1 7, 8 2257
NM_001308200.1 NP_001295129.1 2, 3 505
NM_198567.5 NP_940969.3 4, 5 747
NR_131772.1 4, 5 747
XM_011534553.2 XP_011532855.1 6, 7 6054
XM_011534554.2 XP_011532856.1 6, 7 1926
XM_011534556.2 XP_011532858.1 5, 6 5923
XM_017009454.1 XP_016864943.1 5, 6 1857
XM_017009455.1 XP_016864944.1 6, 7 2167
XM_017009456.1 XP_016864945.1 6, 7 2063
GenBank mRNA BC032390.1 - 2, 3 473
BC037298.1 - 7, 8 2258 4823991
BC066980.1 - 4, 5 740 4821958
BC073880.1 - 2, 3 505
KF151170.1 - 5, 6 1917

Back To Top

More Information


Gene Ontology Categories:

Process(es)

SUMO polymer binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0