This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF517 SIRNA Assays ›
Gene Symbol:
ZNF517
Gene Name:
zinc finger protein 517
Gene Aliases:
-
Chromosome Location:
Chr.8: 144798876 - 144813033 on Build GRCh38
UniGene ID:
Hs.521942
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS139186
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF517
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317936.1 NP_001304865.1 5 1319
NM_213605.2 NP_998770.2 5 1340
NR_134284.1 5 1340
XM_011517014.2 XP_011515316.1 5 1879
XM_011517015.2 XP_011515317.1 5 1321
XM_011517017.2 XP_011515319.1 4 1738
XM_017013384.1 XP_016868873.1 2 1264
GenBank mRNA AK096527.1 - 6 1836
AK097278.1 - 3 2363
AK131440.1 - 5 1340
BC126161.1 - 4 1260 8991877
BC143922.1 - 3 1133

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0