This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM126B SIRNA Assays ›
Gene Symbol:
FAM126B
Gene Name:
family with sequence similarity 126 member B
Gene Aliases:
HYCC2
Chromosome Location:
Chr.2: 200973718 - 201071671 on Build GRCh38
UniGene ID:
Hs.24701
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS138676
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM126B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321618.1 NP_001308547.1 11 1448
NM_001321619.1 NP_001308548.1 12 1659
NM_001321621.1 NP_001308550.1 11 1561
NM_001321622.1 NP_001308551.1 13 1777
NM_001321623.1 NP_001308552.1 13 1714
NM_001321624.1 NP_001308553.1 12 1616
NM_001321625.1 NP_001308554.1 13 1827
NM_001321626.1 NP_001308555.1 13 1847
NM_001321627.1 NP_001308556.1 14 1770
NM_001321628.1 NP_001308557.1 14 1883
NM_001321629.1 NP_001308558.1 13 1602
NM_173822.4 NP_776183.1 12 1546
XM_006712443.3 XP_006712506.1 12 1729
XM_011511002.2 XP_011509304.1 13 1744
XM_017003879.1 XP_016859368.1 14 1925
XM_017003880.1 XP_016859369.1 13 2339
XM_017003881.1 XP_016859370.1 12 1607
XM_017003882.1 XP_016859371.1 14 1826
XM_017003883.1 XP_016859372.1 13 2001
GenBank mRNA AK097111.1 - 5 789
AK315104.1 - 12 1540
AL832234.1 - 11 1405
AL832346.1 - 14 1765
AL833403.1 - 12 1530
BC039295.1 - 12 1544 5267063

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0