This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CCDC150 SIRNA Assays ›
Gene Symbol:
CCDC150
Gene Name:
coiled-coil domain containing 150
Gene Aliases:
-
Chromosome Location:
Chr.2: 196639632 - 196733668 on Build GRCh38
UniGene ID:
Hs.132519
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS138647
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CCDC150
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080539.1 NP_001074008.1 4 659
XM_006712438.3 XP_006712501.1 4 659
XM_011510985.2 XP_011509287.1 3 451
XM_011510988.2 XP_011509290.1 3 430
XM_017003861.1 XP_016859350.1 4 659
XM_017003862.1 XP_016859351.1 4 659
XM_017003863.1 XP_016859352.1 4 659
XM_017003864.1 XP_016859353.1 4 659
XM_017003865.1 XP_016859354.1 3 430
GenBank mRNA AK096979.1 - 4 659
AK303236.1 - 3 425
AK309349.1 - 2 516

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0