This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C20ORF203 SIRNA Assays ›
Gene Symbol:
C20orf203
Gene Name:
chromosome 20 open reading frame 203
Gene Aliases:
-
Chromosome Location:
Chr.20: 32631625 - 32651981 on Build GRCh38
UniGene ID:
Hs.353262
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS138630
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C20orf203
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182584.2 NP_872390.2 3 830
GenBank mRNA AK091025.1 - 3 727
BC105014.1 - 3 606
BC105042.1 - 3 608
GU931819.1 - 4 1140
GU931820.1 - 4 1140

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0